Sulphorhodamine 101-PEO₁₂-propionic acid succinimydyl ester amine-reactive fluorescent dye
Proveedor: Biotium
90100.EA
224
EUR
90100.
Sulphorhodamine 101-PEO₁₂-propionic acid succinimydyl ester amine-reactive fluorescent dye
Sulphorhodamine 101-PEO₁₂-propionic acid succinimydyl ester
SR101-PEO12 SE (Sulforhodamine 101-PEO12-propionic acid succinimydyl ester; Texas Red®-PEO12 SE) has an approximately 47 Angstrom water-soluble PEG spacer that separates the dye from the amine-reactive SE group.
- λEx/λ Em= 583/603nm
- Dark purple solid soluble in DMF or DMSO
- Store at -20°C and protect from light, especially in solution
Texas Red is a registered trademark of Thermo Fisher Scientific.