Polystyrene check beads, for Moxi Flow system
Proveedor: Orflo
ORFLMXA016EA 196 EUR
ORFLMXA016
Polystyrene check beads, for Moxi Flow system
Perlas Bolas de plástico
Moxi Flow check beads are used to verify proper system fluidic (metering), electronic (counts), and optical (fluorescence) performance. Suitable for cassettes Type MF-S and Type MF-M.
  • Formulation: Phosphate-buffered saline, pH 7,4, containing 0,045% sodium azide, surfactant
  • Concentration: 1,1 e+5 beads/ml (5,5 e+4/ml each diameter)
  • Overall dimensions: 22,1 mm³
Store at 2 to 8 °C.
Información para pedidos: Sufficient for 6 tests.
Precaución: For research use only. Product is not for use in diagnostic procedures.