Spectrophotometer, Novaspec® III+
Proveedor: Biochrom
Novaspec® III+
SCLI80-2120-40EA 3850 EUR
SCLI80-2120-40
Spectrophotometer, Novaspec® III+
Espectrofotómetros y Fotómetros Espectrofotómetros UV/Vis
This lightweight instrument is ideal for use in student teaching laboratories. It measures absorbance, % transmission and concentration as well as simple kinetics.
  • Single beam visible spectrophotometer covering 325 to 1100 nm
  • Rugged instrument with no moving parts: Portable and highly reliable
  • Large display allows the user to view results in numerical value as well as a graphical view
  • Up to 72 methods can be stored; methods can be locked using a pin protected password
  • Accepts standard 10 mm pathlength glass or disposable cuvettes
PVC (Print Via Computer) software provides the user with the option to transfer data directly to a PC. Data can be easily stored on a USB memory stick and analysed at a later date.
Información de suministro: Supplied with PVC (Print Via Computer) software.