Multi-parameter meters, handheld, C10xx series
Proveedor: Consort
CONSC1010PEA 695 EUR
CONSC1010P CONSC1020K
Multi-parameter meters, handheld, C10xx series
The C1000 series are basic, complete and accurate bench-top meters. It can measure all common electrochemical parameters such as pH, ORP, conductivity and dissolved oxygen, larger conductivity range, salinity, TDS and measurement storage capability.
  • Easy to use
  • Ideal for schools and small laboratories
  • Large graphical display
  • Stability algorithm with intuitive indicator
  • Interactive LCD screen provides step by step instructions in the language of choice (English, Dutch, French, German)
Bright LCD screen for better readability. A white backlight automatically illuminates when operated on the mains. Stability indicator prompts the user when readings should be taken.