Concrete wedge anchor, DBI-SALA, Saflok®, 3M™
CPSD2100100EA 0 EUR
CPSD2100100
Concrete wedge anchor, DBI-SALA, Saflok®, 3M™
DBI-SALA® Saflok® Concrete wedge anchor for connection into a 19 mm hole in solid concrete.
  • Quick-release button for fast removal with a pivoting easy-grip handle
  • Innovative forged connection ring provides a compatible tie-off point
  • Shielded load cable and chock release cables
Certificaciones: EN 795, Class B