(-)-Perillyl alcohol ≥85%
ABCAAB142452-50EA
698
EUR
ABCAAB142452-50
ABCAAB142452-10
(-)-Perillyl alcohol ≥85%
(-)-Perillyl alcohol
Formula:
C₁₀H₁₆O MW: 152,24 g/mol |
Número MDL:
MFCD00062995 Núm. CAS: 18457-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...