7-Nitroindazole sodium salt ≥98% (by TLC)

Proveedor: Enzo Life Sciences

ALEXALX270200M005EA 37 EUR
ALEXALX270200M005 ALEXALX270200M100
7-Nitroindazole sodium salt ≥98% (by TLC)
7-Nitroindazole sodium salt

A more soluble salt form of 7-nitroindazole. Its higher solubility in artificial cerebrospinal fluid (CSF) permits its use as an inhibitor of nitric oxide synthase (NOS) in rat brain tissue. Provides a useful tool for the study of nitric oxide (NO) activity in the brain.


  • NOS inhibitor


Orange to red solid.


Soluble in 100% ethanol, methanol or DMSO.

Formula: C₇H₄N₃O₂·Na
MW: 162,1 , 23.0 g/mol
Temperatura de almacenaje: Congelador
Núm. CAS: 161467-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR