Cuvette for fluorimeters
Proveedor: Jenway
JENW035121EA 480 EUR
JENW035121
Cuvette for fluorimeters
Glass cuvette for fluorimeters.
  • Four windows and a PTFE lid
Designed for measuring liquids in the visible region.