2,3',5,5'-Tetraclorobifenilo

Proveedor: LGC Standards PROMOCHEM

PROOCIL-PCB-72-CEA 0 EUR
PROOCIL-PCB-72-C
2,3',5,5'-Tetraclorobifenilo
2,3',5,5'-Tetraclorobifenilo
Formula: C₁₂H₆Cl₄
MW: 291,99 g/mol
Núm. CAS: 41464-42-0
UN: 3432
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR