2,2',3,4',5,6,6'-Heptaclorobifenilo

Proveedor: LGC Standards PROMOCHEM

PROOCIL-PCB-188-CEA 304 EUR
PROOCIL-PCB-188-C
2,2',3,4',5,6,6'-Heptaclorobifenilo
2,2',3,4',5,6,6'-Heptaclorobifenilo
Formula: C₁₂H₃Cl₇
MW: 395,32 g/mol
Núm. CAS: 74487-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR