Analogue mini hotplates, basic
Proveedor: Ohaus
Ohaus
OHAU30392012EA 237 EUR
OHAU30392012 OHAU30392033
Analogue mini hotplates, basic
Calentadores Placas calefactoras
These basic mini hotplates are rugged, compact units that heat up to 1000 ml of liquid. Ceramic tops feature a reflective white, chemical resistant top plate that is easy to clean. Bi-metallic thermostat offers reliable temperature control.
  • Compact design saves bench space
  • Ideal for educational laboratories
  • Powerful heater reaches maximum temperature in only minutes (boil 300 ml of water in 18 minutes)
  • Built-in support rod holder with locking knob accepts optional support rod and clamp kit
  • Can be used at 5 to 40 °C (from 20 to max. 80% relative humidity, non condensing)
Basic mini fixed temperature hotplate features an illuminated rocker switch to activate the preset fixed temperature of 500 °C.