Clofibrate ≥98% (por GC)

Proveedor: Enzo Life Sciences
Peligro

ALEXALX270256G001EA 45 EUR
ALEXALX270256G001 ALEXALX270256M500
Clofibrate ≥98% (por GC)
Clofibrate

PPARα activator.


Activates PPARα. Induces cytochrome P450 4A1 and 4A3 which catalise the omega-oxidation of fatty acids and prostaglandins.


Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml).

Formula: C₁₂H₁₅ClO₃
MW: 242,70 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00000615
Núm. CAS: 637-07-0
UN: 3082
ADR: 9,III

Order Now

Resultados test especificaciones

Purity ≥98% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR