VWR®, Filter Tips, Next Generation Refill, Low Retention
Proveedor: VWR
VWR®
732-3350EA 76 EUR
732-3350 732-3351 732-3354 732-3353 732-3347 732-3349
VWR®, Filter Tips, Next Generation Refill, Low Retention
Puntas de pipetas
These aerosol filter tips, with their patented filters made from high molecular weight polyethylene, offer complete protection from sample carry-over. Ideal for PCR and DNA manipulation.
  • Extremely low surface tension significantly reduces sample loss and provides substantially higher reproducibility when working with critical media
  • Easy to use refill system - new pipette tip racks, inserts and transfer cover preventing tip wobbling during refill and keeping tips clean
  • 95% renewable materials - compostable tip base from organic non petroleum resin, carton made from 100% recycled fibres, printing is soy-based vegetable ink
  • Certified human DNA-/RNase-/DNase-free
  • Free from pyrogens, ATP and protease activity
Next Generation is a complete system designed with a low carbon footprint that includes the first ever use of bio-plastic (non petroleum) compostable resin in pipette tip refills. Next Generation’s small footprint extends to the package, with a reclosable box that helps maximise storage space in crowded labs.