NUCLEAR-ID® ≥95% (by HPLC), CELLESTIAL® blue DNA stain

Proveedor: Enzo Life Sciences

ALEXENZ-52406EA 426 EUR
ALEXENZ-52406
NUCLEAR-ID® ≥95% (by HPLC), CELLESTIAL® blue DNA stain
NUCLEAR-ID®

Cell permeable DNA stain that can be used for a wide range of applications


  • Far-red fluorescent specific DNA dye
  • Stable and highly pure
  • Live, permeable and fixed cells can be analyzed
  • No photobleaching effect
  • No RNase treatment is required
  • GFP and FITC compatible
  • UV laser source is not required for excitation
  • Validated for a wide range of cell densitites
  • Quick and easy to use!


The NUCLEAR-ID® Red DNA Stain is a cell permeable dye, designed for use in a range of fluorescence detection technologies, in the discrimination of nucleated cells. It is resistant to photobleaching and is suitable for live-cell staining of nuclei. Also this dye provides a convenient approach for studying the induction and inhibition of cell cycle progression by flow cytometry. Potential applications of this reagent for live-cell studies are in the determination of cellular DNA content and cell cycle distribution, for the detection of variations in growth patterns, for monitoring apoptosis, and for evaluating tumor cell behavior and suppressor gene mechanisms.


Applications:Flow Cytometry, Fluorescent detection

Order Now

Resultados test especificaciones

Excitation maximum: 566nm
Emission maximum: 650nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR