Puromicina diclorhidrato ≥98% (by HPLC)
Proveedor: Enzo Life Sciences
ALEXBML-GR312-0250EA 264 EUR
ALEXBML-GR312-0250 ALEXBML-GR312-0050
Puromicina diclorhidrato ≥98% (by HPLC)
Puromycin dihydrochloride
Inhibits protein synthesis and induces apoptosis.
  • White to off-white solid
  • Soluble in water (50 mg/ml)
Inhibits translation by causing premature release of nascent polypeptide.
Información para pedidos: Shipped at ambient temperature
Formula: C₂₂H₂₉N₇O₅·2HCl
MW: 544,44 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00012691
Núm. CAS: 58-58-2
https://es.vwr.com/stibo/web/std.lang.all/39/50/45403950.jpg