The Q1375 sonicator allows industrial-scale sample volumes to be processed in individual batches or flow-through applications.
- Powerful, durable and customisable
- Set time and amplitude for hands free operation
- Prevents heat buildup in temperature sensitive samples
- Output intensity can be set from 20 to 100%
- Displays duration of sonication, real-time energy monitoring
- Prevents damage to circuitry if a fault occurs and overheating of samples
The booster increases the amplitude of the probe which enables larger volumes to be processed.
Información para pedidos: Q1375 requires a fused, single phase 3-terminal grounding type electrical outlet capable of supplying 50/60 Hz at 220 V or 240 V.
Información de suministro: Supplied with generator, converter, converter cable, power cable and wrench set.