Bead dispenser for 384 wells microtiter or deep well plates
IBSA384-2-999 IBSA384-2-1-20 IBSA384-2-2-10 IBSA384-2-1-10 IBSA382-2-2-20
Bead dispenser for 384 wells microtiter or deep well plates
The 384-well bead dispenser is used to dispense beads in 384-well microtiter plates. Can dispense a wide range of solid quantities in a highly precise and repeatable fashion.
  • Simple, fast and reliable
  • High productivity improvement, 60-150 times
  • Easy cleaning with 70% ethanol or soap
  • Sterilisable and not autoclavable
  • Ergonomic use as it removes tedious repetitive tasks
  • Custom made volume plates are available
Material: ABS+PC, nylon, RVS304, anodised aluminum
* Custom made
Información de suministro: Supplied with converter and flight case.