Knife mills, Grindomix GM 200
Proveedor: Retsch
GRINDOMIX
RETS202540002EA 3550 EUR
RETS202540002 RETS202540001
Knife mills, Grindomix GM 200
Molinos
The GRINDOMIX GM 200 knife mill is the ideal instrument for grinding and homogenising foods and feeds. This can process sample volumes up to 0,7 litres quickly and reproducibly.
  • Efficient size reduction of up to 700 ml feed quantity due to a powerful 1000 W motor
  • Pre- and fine-grinding in one mill: Cutting action in regular mode, grinding by impact in reverse mode, pre-grinding in interval mode
  • Perfect adaptation to application requirements by variable speed from 2000 to 10000 min-1 with an increment of 500 min-1
  • Boost function with speed of 14000 min-1
  • Eight SOP's and four sequences can be stored
  • Touch display with access to My RETSCH web portal with product and application related information
  • Optional gravity lids or volume reduction lids for automatic reduction of the grinding chamber volume
Mills have two sharp, robust blades and a powerful 1000 W motor, ideal for homogenising substances with a high water, oil or fat content as well as for grinding dry, soft and medium-hard products.
A wide selection of lids and containers allows for adaptation of the mill to individual application requirements. This system meets and exceeds all special laboratory and analytical requirements and is a professional device superior to any commercial household mixer.
Información de suministro: Supplied with one litre grinding container made from autoclavable plastic with a standard lid and stainless steel knife. Grinding jars are available in plastic, stainless steel or glass as optional accessories.