Paclitaxel ≥98%
Proveedor: Enzo Life Sciences
Ultra-pure; stabilizes microtubules
Antitumor agent. Stabilizes microtubules and lowers the critical concentration for tubulin polymerization. Reversibly binds to polymerized tubulin. Inhibits protein isoprenylation and induces apoptosis in PC-3 human prostate cancer cells. The checkpoint of mitotic spindle assembly, aberrant activation of cyclin-dependent kinases (CKDs) and the c-Jun N-terminal kinase/stress activated protein kinase (JNK/SAPK) are involved in Paclitaxel-induced apoptosis.
Isolated from the bark of the Pacific Yew tree (Taxus brevifolia)
Formula:
C₄₇H₅₁NO₁₄ MW: 853,91 g/mol |
Número MDL:
MFCD00869953 Núm. CAS: 33069-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...