Ginkgolide B ≥90%
Proveedor: Enzo Life Sciences
ALEXBML-L135-0010EA
59
EUR
ALEXBML-L135-0010
Ginkgolide B ≥90%
Ginkgolide B
PAF receptor ligand
Most potent member of the ginkgolide family of PAF antagonists. Ginkgolide B accelerates PAF hydrolysis by promoting PAF-AH I alpha2 homodimer activity. Inhibits PAF-induced PMNL chemotaxis (IC50=490 nM). Orally effective against PAF-induced bronchospasm.
Formula:
C₂₀H₂₄O₁₀ MW: 424,40 g/mol |
Número MDL:
MFCD05662347 Núm. CAS: 15291-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...