5-Hydroxydecanoate sodium salt ≥98% (by TLC)

Proveedor: Enzo Life Sciences

ALEXBML-KC141-0500EA 338 EUR
ALEXBML-KC141-0500 ALEXBML-KC141-0100
5-Hydroxydecanoate sodium salt ≥98% (by TLC)
5-Hydroxydecanoate sodium salt

Inhibits ATP-sensitive K+ channels.


  • K+ channel blocker


White solid.


Soluble in water (25 mg/ml).

Formula: C₁₀H₁₉O₃·Na
MW: 210,3 g/mol
Número MDL: MFCD00153808
Núm. CAS: 71186-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR