3-Methylcholanthrene ≥97% (by TLC)

Proveedor: Enzo Life Sciences

ALEXBML-GR239-0050EA 327 EUR
ALEXBML-GR239-0050 ALEXBML-GR239-0010
3-Methylcholanthrene ≥97% (by TLC)
3-Methylcholanthrene

Aryl hydrocarbon receptor (AhR) agonist. Inhibits osteoclast formation by inhibiting the expression of receptor activator of NF-κB receptor ligand (RANKL). Suppresses growth factor and JAK-2 expression in mice.


  • Ah receptor ligand


Yellow solid.


Soluble in chloroform or ethyl acetate.

Formula: C₂₁H₁₆
MW: 268,4 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 56-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR