N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED) ≥95% (by NMR)

Proveedor: Enzo Life Sciences

Sinónimos: HBED.HCl.H₂O

ALEXBML-FR113-0050EA 59 EUR
ALEXBML-FR113-0050 ALEXBML-FR113-0250
N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED) ≥95% (by NMR)
N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED)

Potent iron chelator


Iron chelator. Inhibits ferric ion catalyzed formation of hydroxyl radicals. A very potent ligand for the chelation of ferric iron in vivo (active in rat models but inactive in primate models). Inhibits neutrophil apoptosis. An important tool for probing the involvement of hydroxyl radicals in cellular processes.

MW: 424,9 g/mol
Número MDL: MFCD04113603
Núm. CAS: 35369-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR