PD-146176 ≥98% (by HPLC)

Proveedor: Enzo Life Sciences

ALEXBML-EI356-0050EA 454 EUR
ALEXBML-EI356-0050 ALEXBML-EI356-0010
PD-146176 ≥98% (by HPLC)
PD-146176

A potent and selective 15-lipoxygenase inhibitor which lacks significant nonspecific antioxidant properties. It suppresses atherogenesis and limits atherosclerotic lesion development in the rabbit. It reduces oxidant stress-induced apoptosis in endothelial cells and inhibits proliferation in 15-LO overexpressing PC3 cells.


  • Lipoxygenase inhibitor


A potent and selective 15-lipoxygenase inhibitor which lacks significant nonspecific antioxidant properties. It suppresses atherogenesis and limits atherosclerotic lesion development in the rabbit. It reduces oxidant stress-induced apoptosis in endothelial cells and inhibits proliferation in 15-LO overexpressing PC3 cells.

Formula: C₁₅H₁₁NS
MW: 237,3 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD05664738
Núm. CAS: 4079-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR