URB-602 ≥98% (by TLC)
Proveedor: Enzo Life Sciences
ALEXBML-EI348-0050EA
310
EUR
ALEXBML-EI348-0050
ALEXBML-EI348-0010
URB-602 ≥98% (by TLC)
URB-602
Selectively inhibits monoacylglycerol (MAG) lipase (IC50=28µM) via a noncompetitive mechanism. Does not inhibit FAAH, DAG lipase or COX-2. URB-602 selectively raises levels of 2-arachidonoylglycerol with no change in anandamide concentrationsin cells and tissues. In the same way anandamide levels were markely elevated by the FAAH inhibitor URB597 at 1µM.
- Monoacylglycerol lipase inhibitor
Off-white solid.
Soluble in DMSO (30 mg/ml) or ethanol (5 mg/ml warm).
Formula:
C₁₉H₂₁NO₂ MW: 295,4 Pto. de fusión: 122…123 °C Temperatura de almacenaje: Ambiente |
Núm. CAS:
565460-15-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...