1-Aminobenzotriazol

Proveedor: Enzo Life Sciences

ALEXBML-EI259-0100EA 142 EUR
ALEXBML-EI259-0100
1-Aminobenzotriazol
1-Aminobenzotriazole

Cytochrome P450 inhibitor


Suicide substrate for cytochrome P-450. Inhibits oxidative drug metabolism in vivo. Blocks omega-oxidation of arachidonic acid but not epoxidation.

Formula: C₆H₆N₄
MW: 134,14 g/mol
Número MDL: MFCD00132902
Núm. CAS: 1614-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR