Bisindolylmaleimide III ≥98%

Proveedor: Enzo Life Sciences

ALEXALX270051M001EA 69 EUR
ALEXALX270051M001 ALEXALX270051M005
Bisindolylmaleimide III ≥98%
Bisindolylmaleimide III

PKC inhibitor


Potent and selective inhibitor of protein kinase C (PKC).

Formula: C₂₃H₂₀N₄O₂
MW: 384,4 g/mol
Núm. CAS: 137592-43-9
https://es.vwr.com/stibo/web/std.lang.all/74/64/45347464.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR