Kanamicina sulfato

Proveedor: Enzo Life Sciences

ALEXALX380275G005EA 126 EUR
ALEXALX380275G005
Kanamicina sulfato
Kanamicin sulfato

Antibiotic. Apoptosis inducer.


Isolated from Streptomyces kanamyceticus.


Aminoglycoside antibiotic. Binds to the 70S ribosomal subunit. Inhibits translocation and elicits miscoding. Interaction with ribosomes may be prevented by aminoglycoside modifying enzymes. Effective against Gram-negative and Gram-positive bacteria and mycoplasma.

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582,6 g/mol
Número MDL: MFCD00070253
Núm. CAS: 25389-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR