Vancomycin hydrochloride (de Streptomyces lysosup)
Proveedor: Enzo Life Sciences
ALEXALX380279M250EA
91
EUR
ALEXALX380279M250
ALEXALX380279G001
Vancomycin hydrochloride (de Streptomyces lysosup)
Vancomycin hydrochloride
Glycopeptide antibiotic. Interferes with cell wall synthesis. Effective against Gram-positive bacteria.
White to light brown powder
Soluble in water (50 mg/mL)
Envase: Shipping temperature: Ambient
Formula:
C₆₆H₇₅Cl₂N₉O₂₄·HCl MW: 1485,71 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD03613611 Núm. CAS: 1404-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...