Nile red ≥95% (by HPLC), grado ultrapuro

Proveedor: Enzo Life Sciences

ALEXENZ-52551EA 121 EUR
ALEXENZ-52551
Nile red ≥95% (by HPLC), grado ultrapuro
Nile red

Lipid dye


Nile red is a lipophilic probe that becomes intensely fluorescent in a hydrophobic environment, while it has minimal fluorescence in aqueous media. It is an excellent vital stain for the detection of intracellular lipid droplets by fluorescence microscopy, in cells such as adipocyes. Flow cytometry methods using a single staining with Nile red and double staining with Nile red and anti-CD3 monoclonal antibody have been employed to monitor peripheral leukocyte and lymphocyte phospholipidosis. Nile Red has also been used for rapid staining of proteins in SDS-polyacrylamide gel electrophoresis. Wavelength Maxima: Excitation 552 nm, Emission 636 nm.

Formula: C₂₀H₁₈N₂O₂
MW: 318,38 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00011639
Núm. CAS: 7385-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR