Cathepsin K assay buffer
Proveedor: Enzo Life Sciences
ALEXBML-KI432-0020EA 40 EUR
ALEXBML-KI432-0020
Cathepsin K assay buffer
Tampones Tampones de ensayo
50mM NaOAc, pH 5.5, 2.5mM EDTA, 1mM DTT and 0.01% Triton X-100®