Anti-VDAC Rabbit Polyclonal Antibody
Proveedor: Axxora
ALEXALX210785C100EA 616 EUR
ALEXALX210785C100
Anti-VDAC Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: VDAC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Monkey