Powder dispenser for 384 wells microtiter or deep well plates
IBSA384-1-10 IBSA384-1-999 IBSA384-1-50 IBSA384-1-20
Powder dispenser for 384 wells microtiter or deep well plates
The 384-well powder dispenser is used to dispense the specific amount of powder in a 384-well microtiter plate. Can dispense a wide range of solid quantities in a highly precise and repeatable fashion.
  • Simple, fast and reliable
  • High productivity improvement 60-150 x
  • Easy cleaning with 70% ethanol or soap
  • Sterilisable and not autoclavable
  • Ergonomic use as it removes tedious repetitive tasks
  • Custom made volume plates are available
Material: ABS+PC, nylon, RVS304, anodised aluminum
* Customised capacity (µl)
Información de suministro: Supplied converter and flight case.