Estereomicroscopio, VisiScope® SZT350-6
630-2721EA 1320 EUR
VWRI630-2721
Estereomicroscopio, VisiScope® SZT350-6
Trinocular stereo microscope with extended zoom ratio for processing bigger samples.
  • Greenough optical system
  • Pillar stand with rack, pinion system and ultra-flat base
  • Adjustable brightness for outstanding images
Used for observations in advanced teaching, laboratory and industrial applications.
Información para pedidos: A cleaning and maintenance set is available.