Anti-PTPMT1 Rabbit Polyclonal Antibody
Proveedor: ProSci
PRSI48-194EA 480 EUR
PRSI48-194
Anti-PTPMT1 Rabbit Polyclonal Antibody
Provisional protein phosphatase.
PTPMT1 antibody can be used in Western Blot starting at 1 μg/mL, and immunohistochemistry starting at 5 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PTPMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human