The statistical printer is a bidirectional thermal printer that can be used with laboratory balances. It can print weight values and, when requested, a full statistical analysis of the values including: Number of values, mean value, sum, minimum, maximum, standard deviation and more. The printer can also record and print date/time and calibration details in compliance with GLP.
- Count and total of all measurements, minimum, maximum values, range (max-min)
- Average value, standard deviation
- GLP record keeping of balance calibration with date/time print
- Available units: mg, g, kg, pcs
- Languages: English, French, German
Información de suministro: Supplied with connection cable for direct connection to a balance.
Precaución: Printer can not be used with moisture analysers or balances with a graphic display/touchscreen.