Esta buscando: Analizadores de humedad
22  results were found
SearchResultCount:"22"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Proveedor: VWR Collection
Descripción: Robustos analizadores de humedad compactos con una gran pantalla gráfica de matriz de puntos retroiluminada para la determinación de la humedad de las...
Proveedor: VWR Collection
Descripción: Moisture analysers with large colour touch screen display give a user-friendly access to advanced applications and internal databases. The user has an...
New Product
Numero del catalogo: (620-1956)
Proveedor: VWR Collection
Descripción: The RM 10 allows detection of moisture behind ceramic tiles and/or various wall or floor coverings, in wood down to a depth of 3 cm and in concrete or...
UOM: 1 * 1 UN
Numero del catalogo: (611-2888)
Proveedor: Sartorius Balances
Descripción: Compact moisture analyser giving fast repeatable results with an improved user interface.
UOM: 1 * 1 UN
Numero del catalogo: (611-2887)
Proveedor: Sartorius Balances
Descripción: Easy to use, efficient, compact moisture analysers to give fast repeatable results with an improved user interface.
UOM: 1 * 1 UN
Proveedor: Sartorius Balances
Descripción: The MA100 is an accurate and configurable model with a user friendly keyboard and software prompts to easily access all the modes of operation and dat...
Numero del catalogo: (611-4357)
Proveedor: Sartorius Balances
Descripción: This infrared moisture analyser is designed for use in daily routine in-process control.
UOM: 1 * 1 UN
Numero del catalogo: (SIMELI9010)
Proveedor: Industrial Physics
Descripción: Encounter Plus is a non-destructive moisture meter designed fo the inspection in numerous industries.
UOM: 1 * 1 UN
Numero del catalogo: (SIMELI0004)
Proveedor: Industrial Physics
Descripción: mini LIGNO duo is a wood moisture meter that provides four different base-wood groups, eight building materials, such as gypsum or screed.
UOM: 1 * 1 UN
Proveedor: OHAUS
Descripción: Proporciona resultados precisos y reproducibles con una legibilidad del 0,1% o del 0,05%.
Numero del catalogo: (SIMELI9011)
Proveedor: Industrial Physics
Descripción: Tramex is a non-destructive instrument for measuring moisture levels in concrete and sand-cement floors.
UOM: 1 * 1 UN
Numero del catalogo: (SIMELI0006)
Proveedor: Industrial Physics
Descripción: The mini LIGNO classic is a wood moisture meter that offers the choice between four different base-wood groups.
UOM: 1 * 1 UN
Proveedor: OHAUS
Descripción: Moisture analysers simplify complex moisture analysis applications. Designed to increase precision and efficiency in the laboratory, they feature a pr...
Numero del catalogo: (SIMELI0005)
Proveedor: Industrial Physics
Descripción: mini LIGNO smart is a wood moisture meter that offers the freely selectable choice between up to 16 different species of wood or building materials wi...
UOM: 1 * 1 UN
Numero del catalogo: (SIMELI9012)
Proveedor: Industrial Physics
Descripción: Skipper Plus is a non-destructive moisture detector designed for locating damp in wood or laminated plastics such as polyester.
UOM: 1 * 1 UN
Proveedor: Industrial Physics
Descripción: profi LIGNO are moisture meters specifically designed for wood and building materials such as screed or gypsum.
Consulta de precio
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta. Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta.
Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
El producto(s) marcados con este símbolo están descatalogados - se venden hasta acabar stock. Pueden encontrar alternativas buscando con el número de catálogo VWR listado arriba. Si necesita más ayuda, por favor llame al Servicio de atención al cliente de VWR al 902.222.897.
1 - 16 of 22
no targeter for Bottom