Half masks, Profile 60
Proveedor: Scott Safety
SCTS2016138EA 37.55 EUR
SCTS2016138
Half masks, Profile 60
Profile 60 is a twin filter premium half mask with liquid silicone rubber (LSR) facepiece, combining the comfort and flexibilty of silicone with durability and a long service life.
  • LSR injection moulding prevents silicone vapour release
  • Wide and flat faceseal to achieve an efficient and comfortable fit
  • Slide-through harness provides fast and easy fitting and a convenient rest position
Suitable for use in paint-spray areas
Certificaciones: EN 140