Filters, ProFlex, for Aviva half masks
Proveedor: Scott Safety
SCTS8005175EA 907 EUR
SCTS8005175 SCTS8005174 SCTS8005173 SCTS8005172
Filters, ProFlex, for Aviva half masks
These Pro2Flex are low cost particulate filters with negative leak-check feature. Easy to install.