Senescence detection kit, Calbiochem®
Proveedor: Merck Millipore (Oncogene)
Calbiochem®
QIA117-1EA 687 EUR
QIA117-1
Senescence detection kit, Calbiochem®
Ensayos Ensayos celulares
The Calbiochem® Senescence Detection Kit is useful for the histochemical detection of senescent cells. The failure of cells to become senescent may lead to tumour progression. The mechanism of this is believed to be the increased lysosomal content of senescent cells resulting in an increase of the lysosomal enzyme β-galactosidase.
Información de suministro: The kit includes fixative solution, X-Gal, staining solution, staining supplement, and a user protocol.