Anti-EPCAM Monoclonal Antibody [clone: EGP40/1555R]

Proveedor: Abnova
ABNORAB00340EA 492 EUR
ABNORAB00340
Anti-EPCAM Monoclonal Antibody [clone: EGP40/1555R]
Anticuerpos
Recombinant rabbit monoclonal antibody raised against human EPCAM.

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: EGP40/1555R
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR