NAD/NADH ratio assay kit (red)

Proveedor: Abnova
ABNOKA2523EA 612 EUR
ABNOKA2523
NAD/NADH ratio assay kit (red)
NAD/NADH ratio assay kit (red) provides a convenient method for sensitive detection of NAD, NADH and their ratio in solution and cell extracts.

  • Detection range: 540/590 nm
  • Sample volume: 50 µl
  • Storage temperature: −20 °C
  • Absorbance: 340 nm

This kit is easily adapted to automation without a separation step and formulated to have minimal hands-on time. No special requirement is needed for waste disposal.

Información de suministro: The kit contains 1×96-well plate pre-coated with anti-NT-proBNP antibody, lyophilised standards (2 tubes), wash buffer (20×50 ml), diluent buffer (10 ml), HRP conjugated anti-NT-proBNP antibody (RTU) (6 ml), stop solution (6 ml), TMB chromogenic reagent A (6 ml), TMB chromogenic reagent B (6 ml) and 2×plate sealer.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR