High visibility waistcoat, flame retardant, Vine
Proveedor: Sioen
SIOE9052APEFYXLEA 14.5 EUR
SIOE9052APEFYXL SIOE9052APEFFY1L SIOE9052APEFFY1M SIOE9052APEFY2XL SIOE9052APEFY3XL
High visibility waistcoat, flame retardant, Vine
Ropa de trabajo
Waistcoat made of 100% FR polyester mesh, with reflective tape.
  • Stitched seams
  • V-neck
  • With zip
Back length: 70 cm
Weight: 120 g/m²
Certificaciones: EN 471 + A1 (2-2)
GO/RT 3279 (only for hi-vis orange)