Chamber furnaces, N 11/HR
Proveedor: Nabertherm
NABEN02R0NBNEEA 3990 EUR
NABEN02R0NBNE
Chamber furnaces, N 11/HR
Chamber furnaces for annealing, hardening and brazing.
  • Three-sided heating from both sides and the bottom
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Bottom heating protected by heat-resistant SiC plate
  • Low energy consumption due to multi-layer insulation
  • Parallel guided downward swinging door (user protected from heat radiation)
  • Gas spring dampers ensure easy door opening and closing
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive