Helix pomatia ß-Glucuronidase/aryl sulphatase
Proveedor: Merck
1.04114.0002EA 136 EUR
1.04114.0002
Helix pomatia ß-Glucuronidase/aryl sulphatase
Enzimas
Stabilised aqueous solution for biochemistry EC 3.2.1.31 + EC 3.1.6.1.