Emergency body shower, wall-mounted
Proveedor: Broen Lab
BROE17081.009EA 0 EUR
BROE17081.009
Emergency body shower, wall-mounted
Body shower with self-draining head and pull rod. In brass with chemical resistant polycoating.
  • Quick mounting system
  • Shower head with improved spray pattern
  • Self-draining, no dripping
  • Pre-mounted valve for body shower
  • Supporting wall flange can be used for mounting
Flow rate: ~80 l/min at 1 bar flow pressure
Connection: G½“ male
Depth: 600 mm
Weight: ~3 kg
Certificaciones: EN 15154-1 /-2