Drain covers, PIG® DrainBlocker® Drive-Over
Proveedor: New Pig
NEWPPLR303EA 520.75 EUR
NEWPPLR303 NEWPPLR302
Drain covers, PIG® DrainBlocker® Drive-Over
This tough storm drain cover with its patented design is the only one that can handle drive-overs - even from heavy trucks.
  • Eliminates the need to frequently deploy and remove a temporary cover or permanently seal your drains
  • Tough, patented-rubber HogHyde Skin stands up to punctures, tearing and abrasion, even under heavy vehicle traffic from tankers, trucks or forklifts
  • Super-sealing urethane bottom layer conforms to uneven surfaces and tightly seals drain openings to block liquid
  • Withstands UV rays, infrared light and all weather conditions; keep outdoors as long as needed without worrying about degradation
  • Polyurethane and rubber materials resist water, oils and many chemicals
Ideal for long-term drain protection where liquid transfers, washdowns or other leak- and spill-prone jobs are common.
Colour: Black (top) and blue (bottom)
Información de suministro: Includes a nylon storage bag for easy transportation and fast spill response.