Anti-CD40 Mouse Monoclonal Antibody (APC) [clone: HI40a]

Proveedor: Abnova
ABNOMAB13895EA 471 EUR
ABNOMAB13895
Anti-CD40 Mouse Monoclonal Antibody (APC) [clone: HI40a]
Anticuerpos
Mouse monoclonal antibody raised against human CD40.

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: HI40a
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR