These data loggers are available with or without hygrometer, recording temperature or temperature and humidity data for 30 days. Hourly data recorded for current 7 days, then maximum and minimum temperatures or temperatures and humidity are recorded for next 23 days.
- Monitor critical environments without a physical connection; maximum signal distance is approx. 40 metres
- Alarm sounds if temperature falls outside of the set parameters
- Data viewable as a chart or graph, or on a calendar; data from up to six data loggers can be plotted on one graph for direct comparison
- Indicators include WBGT (Wet Bulb Global Temperature), Heat, Mold and Comfort indices as well as memory full, active alarm state and low battery
- Features all-weather housing with internal temperature sensor and humidity sensor
Certificaciones: Conforms to ROHS compliance and CE marked.
Información de suministro: Supplied with Certificate of Compliance indicating accuracy for four temperature points or four temperature and three humidity points, two AAA batteries, and multi-language (English, French, German, Italian, Portuguese and Spanish) instructions.