VWR®, Absorbents - Mats, Rolls and Socks for Oil Spills
Proveedor: VWR
VWR®
121-0143EA 40.25 EUR
121-0143 121-0140 121-0137
VWR®, Absorbents - Mats, Rolls and Socks for Oil Spills
Materiales para contención de derrames
VWR® absorbents and spill-containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.
  • Will only absorb oil and petroleum-based liquids
  • Will not absorb water-based liquids
  • Can be used outdoors as they will not absorb water
Colour: White